accessibility menu, dialog, popup

UserName

CAS RN 156-41-2

NH3Cl

CAS RN 156-41-2

IUPAC Name: 2-(4-chlorophenyl)ethan-1-aminium
Synonyms: 2-(4-chlorophenyl)ethanaminium
Molecular Weight (g/mol): 156.63
Molecular Formula: C8H11ClN
InChi Key: SRXFXCKTIGELTI-UHFFFAOYSA-O
SMILES: [NH3+]CCC1=CC=C(Cl)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)