CAS RN 155-30-6
CAS RN 155-30-6
IUPAC Name:
(3R,5R,6S)-2-(hydroxymethyl)-6-(methylsulfanyl)oxane-3,4,5-triol
Synonyms:
(3R,5R,6S)-2-(hydroxymethyl)-6-(methylsulfanyl)oxane-3,4,5-triol
Molecular Weight (g/mol):
210.24
Molecular Formula:
C7H14O5S
InChi Key:
LZFNFLTVAMOOPJ-XMAYFYEJSA-N
SMILES:
CS[C@@H]1OC(CO)[C@H](O)C(O)[C@H]1O
1
–
1
of
1
results
1
Promotions Available
Methyl β-D-thiogalactoside, 98%, Thermo Scientific™
Methyl β-D-thiogalactoside, 98%, Thermo Scientific™
CAS: 155-30-6 Molecular Formula: C7H14O5S Molecular Weight (g/mol): 210.25 InChI Key: LZFNFLTVAMOOPJ-PZRMXXKTSA-N Synonym: methyl-beta-d-thiogalactoside,methyl-1-thio-beta-d-galactopyranoside,methyl 1-thio-beta-d-galactopyranoside,2r,3r,4s,5r,6s-2-hydroxymethyl-6-methylsulfanyl oxane-3,4,5-triol,b-d-thiomethylgalactoside,methyl b-d-thiogalactoside,galactose-derived compound 19,methyl beta-d-thio-galactopyranoside,methyl 1-thio-??-d-galactopyranoside,1-s-methyl-1-thio-beta-d-galactopyranose PubChem CID: 10036019 IUPAC Name: (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methylsulfanyloxane-3,4,5-triol SMILES: CSC1C(C(C(C(O1)CO)O)O)O