accessibility menu, dialog, popup

UserName

CAS RN 15486-19-8

CH3OOOHO(E)

CAS RN 15486-19-8

IUPAC Name: (2E)-3-[4-(acetyloxy)phenyl]prop-2-enoic acid
Synonyms: (2E)-3-[4-(acetyloxy)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 206.2
Molecular Formula: C11H10O4
InChi Key: BYHBHNKBISXCEP-QPJJXVBHSA-N
SMILES: CC(=O)OC1=CC=C(\C=C\C(O)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results