accessibility menu, dialog, popup

UserName

FFFONN

CAS RN 1546-79-8

IUPAC Name: 2,2,2-trifluoro-1-(1H-imidazol-1-yl)ethan-1-one
Synonyms: 2,2,2-trifluoro-1-(imidazol-1-yl)ethanone
Molecular Weight (g/mol): 164.09
Molecular Formula: C5H3F3N2O
InChi Key: SINBGNJPYWNUQI-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)N1C=CN=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results