accessibility menu, dialog, popup

UserName

CAS RN 15364-94-0

H2OH2OH2OH2OH2OH2OMnOClOOOOClOOO

CAS RN 15364-94-0

IUPAC Name: manganese(2+) hexahydrate diperchlorate
Synonyms: manganese(2+) hexahydrate diperchlorate
Molecular Weight (g/mol): 361.92
Molecular Formula: Cl2H12MnO14
InChi Key: AIFGVOMWPFMOCN-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.[Mn++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Physical Form 
  • (5)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)