CAS RN 153277-35-1
CAS RN 153277-35-1
IUPAC Name:
9,10-dioxo-9,10-dihydroanthracene-2-sulfonate
Synonyms:
9,10-dioxoanthracene-2-sulfonate
Molecular Weight (g/mol):
287.27
Molecular Formula:
C14H7O5S
InChi Key:
MMNWSHJJPDXKCH-UHFFFAOYSA-M
SMILES:
[O-]S(=O)(=O)C1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1
1
–
1
of
1
results
1
9,10-Anthraquinone-2-sulfonic acid sodium salt hydrate, 97% (dry wt.), water ca 4-6%
CAS: 153277-35-1 Molecular Formula: C14H7NaO5S Molecular Weight (g/mol): 310.255 MDL Number: MFCD00149068 InChI Key: GGCZERPQGJTIQP-UHFFFAOYSA-M Synonym: sodium anthraquinone-2-sulfonate,anthraquinone-2-sulfonic acid sodium salt,sodium 2-anthraquinonesulfonate,unii-511h72sdvx,sodium 9,10-dioxo-9,10-dihydroanthracene-2-sulfonate,2-anthracenesulfonic acid, 9,10-dihydro-9,10-dioxo-, sodium salt,sodium 2-anthrachinonesulphonate,2-anthraquinone sodium sulfonate,2-anthraquinonesulfonic acid sodium salt,sodium-2-anthrachinonesulphonate PubChem CID: 23661981 IUPAC Name: sodium;9,10-dioxoanthracene-2-sulfonate SMILES: C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)S(=O)(=O)[O-].[Na+]