accessibility menu, dialog, popup

UserName

CH3OOCH3OOCH3OODyH2OH2OH2OH2O

CAS RN 15280-55-4

IUPAC Name: dysprosium(3+) triacetate
Synonyms: dysprosium(3+) triacetate
Molecular Weight (g/mol): 339.63
Molecular Formula: C6H9DyO6
InChi Key: HKCSKROOVZMKSO-UHFFFAOYSA-K
SMILES: [Dy+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Grade 
  • (4)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)