CAS RN 152362-01-1
CAS RN 152362-01-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Chloro-4-methylnicotinamide, TRC
CAS: 152362-01-1 Molecular Formula: C7H7N2OCl Molecular Weight (g/mol): 170.6 Synonym: 2-Chloro-4-methyl-3-pyridinecarboxamide,2-Chloro-4-methyl-nicotinamide IUPAC Name: 2-chloro-4-methylpyridine-3-carboxamide SMILES: Cc1ccnc(Cl)c1C(=O)N