accessibility menu, dialog, popup

UserName

CAS RN 1522-46-9

H3COOCH3H3CH3CO

CAS RN 1522-46-9

IUPAC Name: ethyl 2-acetyl-3-methylbutanoate
Synonyms: ethyl 2-acetyl-3-methylbutanoate
Molecular Weight (g/mol): 172.22
Molecular Formula: C9H16O3
InChi Key: DMIFFKCVURTPTG-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C(C)C)C(C)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results