accessibility menu, dialog, popup

UserName

CAS RN 1504-58-1

HO

CAS RN 1504-58-1

IUPAC Name: 3-phenylprop-2-yn-1-ol
Synonyms: 3-phenylprop-2-yn-1-ol
Molecular Weight (g/mol): 132.16
Molecular Formula: C9H8O
InChi Key: NITUNGCLDSFVDL-UHFFFAOYSA-N
SMILES: OCC#CC1=CC=CC=C1
Boiling Point 
  • (6)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (6)
  • (2)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)