CAS RN 1500-85-2
IUPAC Name:
7H-pyrrolo[2,3-d]pyrimidin-4-amine
Synonyms:
7-deazaadenine
Molecular Weight (g/mol):
134.14
Molecular Formula:
C6H6N4
InChi Key:
PEHVGBZKEYRQSX-UHFFFAOYSA-N
SMILES:
NC1=C2C=CNC2=NC=N1
1
–
1
of
1
results
1
6-Amino-7-deazapurine, 97%, Thermo Scientific™
CAS: 1500-85-2 Molecular Formula: C6H6N4 Molecular Weight (g/mol): 134.14 MDL Number: MFCD01686848 InChI Key: PEHVGBZKEYRQSX-UHFFFAOYSA-N Synonym: 4-amino-7h-pyrrolo 2,3-d pyrimidine,7h-pyrrolo 2,3-d pyrimidin-4-amine,7-deazaadenine,7h-pyrrolo 2,3-d pyrimidin-4-ylamine,4-aminopyrrolo 2,3-d pyrimidine,1h-pyrrolo 2,3-d pyrimidin-4-amine,7h-pyrrolo 2,3-d pyrimidine, 4-amino,pubchem14980 PubChem CID: 5359620 IUPAC Name: 7H-pyrrolo[2,3-d]pyrimidin-4-amine SMILES: NC1=C2C=CNC2=NC=N1