accessibility menu, dialog, popup

UserName

CAS RN 14972-70-4

RhClClClClClClNaNaNaH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2O

CAS RN 14972-70-4

IUPAC Name: rhodium(3+) trisodium hexachloride
Synonyms: rhodium(3+) trisodium hexachloride
Molecular Weight (g/mol): 384.57
Molecular Formula: Cl6Na3Rh
InChi Key: YDEXHLGYVJSKTN-UHFFFAOYSA-H
SMILES: [Na+].[Na+].[Na+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Rh+3]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results