CAS RN 14933-09-6
CAS RN 14933-09-6
IUPAC Name:
3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate
Synonyms:
myristyl sultaine
Molecular Weight (g/mol):
363.6
Molecular Formula:
C19H41NO3S
InChi Key:
BHATUINFZWUDIX-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O
1
–
1
of
1
results
1
Promotions Available
3-(N,N-Dimethylmyristylammonio)propanesulfonate, 97%
3-(N,N-Dimethylmyristylammonio)propanesulfonate, 97%
CAS: 14933-09-6 Molecular Formula: C19H41NO3S Molecular Weight (g/mol): 363.61 MDL Number: MFCD00036910 InChI Key: BHATUINFZWUDIX-UHFFFAOYSA-N Synonym: myristyl sulfobetaine,zwittergent 3-14,n-tetradecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate,3-dimethyl tetradecyl ammonio propane-1-sulfonate,3-n,n-dimethylmyristylammonio propanesulfonate,dimethyl 3-sulphonatopropyl tetradecylammonium,zwittergent™ 3-14 detergent,3-myristyldimethylammonio propanesulfonate,3-n,n-dimethyltetradecylammonio propanesulfonate PubChem CID: 84705 ChEBI: CHEBI:10129 IUPAC Name: 3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate SMILES: CCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]