accessibility menu, dialog, popup

UserName

CAS RN 149169-34-6

SH2NOFFF

CAS RN 149169-34-6

IUPAC Name: 4-(trifluoromethoxy)benzene-1-carbothioamide
Synonyms: 4-(trifluoromethoxy)benzenecarbothioamide
Molecular Weight (g/mol): 221.2
Molecular Formula: C8H6F3NOS
InChi Key: GHQSKHMIUWHKHO-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results