CAS RN 148476-30-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-Mesityl-2-oxo-1-oxaspiro[4.4]non-3-en-4-ol(Spiromesifen Metabolite M01), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-Mesityl-2-oxo-1-oxaspiro[4.4]non-3-en-4-ol(Spiromesifen Metabolite M01), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.