CAS RN 1483-28-9
IUPAC Name:
2,5-dimethoxybenzene-1-sulfonyl chloride
Synonyms:
2,5-dimethoxybenzenesulfonyl chloride
Molecular Weight (g/mol):
236.67
Molecular Formula:
C8H9ClO4S
InChi Key:
SHELADVIRCCTFN-UHFFFAOYSA-N
SMILES:
COC1=CC=C(OC)C(=C1)S(Cl)(=O)=O
1
–
1
of
1
results
1
2,5-Dimethoxybenzenesulfonyl chloride, 98%, Thermo Scientific™
CAS: 1483-28-9 Molecular Formula: C8H9ClO4S Molecular Weight (g/mol): 236.67 MDL Number: MFCD00024872 InChI Key: SHELADVIRCCTFN-UHFFFAOYSA-N Synonym: 2,5-dimethoxybenzene-1-sulfonyl chloride,2,5-dimethoxybenzenesulphonyl chloride,benzenesulfonyl chloride, 2,5-dimethoxy,2,5-dimethoxyphenyl chlorosulfone,2,5-dimethoxybenzenesulphonylchloride,2,5-dimethoxy-benzenesulfonyl chloride,2,5-dimethoxybenzenesulfonylchloride,asischem d48918,pubchem5126,timtec-bb sbb003243 PubChem CID: 137025 IUPAC Name: 2,5-dimethoxybenzenesulfonyl chloride SMILES: COC1=CC=C(OC)C(=C1)S(Cl)(=O)=O