accessibility menu, dialog, popup

UserName

CAS RN 14813-01-5

OHN

CAS RN 14813-01-5

IUPAC Name: 1-benzylpiperidin-3-ol
Synonyms: 1-benzylpiperidin-3-ol
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: UTTCOAGPVHRUFO-UHFFFAOYNA-N
SMILES: OC1CCCN(CC2=CC=CC=C2)C1
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)