accessibility menu, dialog, popup

UserName

CAS RN 14794-32-2

H3COOOCl

CAS RN 14794-32-2

IUPAC Name: ethyl 7-chloro-7-oxoheptanoate
Synonyms: ethyl 7-chloro-7-oxoheptanoate
Molecular Weight (g/mol): 206.67
Molecular Formula: C9H15ClO3
InChi Key: NYZNYLOIPHDFKS-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCCC(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results