CAS RN 147624-13-3
IUPAC Name:
3-fluoro-2-methylbenzaldehyde
Synonyms:
3-fluoro-2-methylbenzaldehyde
Molecular Weight (g/mol):
138.14
Molecular Formula:
C8H7FO
InChi Key:
HSUCYMJBFHBMTB-UHFFFAOYSA-N
SMILES:
CC1=C(F)C=CC=C1C=O
1
–
1
of
1
results
1
3-Fluoro-2-methylbenzaldehyde, 98%, Thermo Scientific™
CAS: 147624-13-3 Molecular Formula: C8H7FO Molecular Weight (g/mol): 138.141 MDL Number: MFCD00075256 InChI Key: HSUCYMJBFHBMTB-UHFFFAOYSA-N Synonym: 2-methyl-3-fluorobenzaldehyde,3-fluoro-2-methyl-benzaldehyde,benzaldehyde, 3-fluoro-2-methyl,pubchem1453,acmc-1cfbd,intermediates-zcf02128,ksc494i0h,attercop-chm at111499,3-fluoro-2-methylbenzaldehyde PubChem CID: 2779235 IUPAC Name: 3-fluoro-2-methylbenzaldehyde SMILES: CC1=C(C=CC=C1F)C=O