accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OH2OH2OOClOOOOClOOOOClOOOEr

CAS RN 14692-15-0

IUPAC Name: erbium(3+) hexahydrate triperchlorate
Synonyms: erbium(3+) hexahydrate triperchlorate
Molecular Weight (g/mol): 573.69
Molecular Formula: Cl3ErH12O18
InChi Key: JZRNHNTYRRYOCR-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.[Er+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)