accessibility menu, dialog, popup

UserName

CAS RN 14683-47-7

CH3HN

CAS RN 14683-47-7

IUPAC Name: (diphenylmethyl)(methyl)amine
Synonyms: (diphenylmethyl)(methyl)amine
Molecular Weight (g/mol): 197.28
Molecular Formula: C14H15N
InChi Key: SHDMMLFAFLZUEV-UHFFFAOYSA-N
SMILES: CNC(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results