accessibility menu, dialog, popup

UserName

CAS RN 1467-79-4

CH3NCH3N

CAS RN 1467-79-4

IUPAC Name: cyanodimethylamine
Synonyms: dimethyl cyanamide
Molecular Weight (g/mol): 70.1
Molecular Formula: C3H6N2
InChi Key: OAGOUCJGXNLJNL-UHFFFAOYSA-N
SMILES: CN(C)C#N
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)