CAS RN 1465-16-3
CAS RN 1465-16-3
IUPAC Name:
3-[2-(dimethylamino)ethyl]-1-methyl-1H-indol-4-ol
Synonyms:
1-methylpsilocin
Molecular Weight (g/mol):
218.3
Molecular Formula:
C13H18N2O
InChi Key:
MZZRFEIDRWKTKJ-UHFFFAOYSA-N
SMILES:
CN(C)CCC1=CN(C)C2=CC=CC(O)=C12
1
–
1
of
1
results
1
Tocris Bioscience™ 1-Methylpsilocin
CAS: 1465-16-3 Molecular Formula: C13H18N2O