accessibility menu, dialog, popup

UserName

CAS RN 14643-87-9

ZnOOH2COOH2C

CAS RN 14643-87-9

IUPAC Name: zinc(2+) bis(prop-2-enoate)
Synonyms: zinc(2+) diacrylate
Molecular Weight (g/mol): 207.49
Molecular Formula: C6H6O4Zn
InChi Key: XKMZOFXGLBYJLS-UHFFFAOYSA-L
SMILES: [Zn++].[O-]C(=O)C=C.[O-]C(=O)C=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results