accessibility menu, dialog, popup

UserName

CAS RN 146140-95-6

IUPAC Name: [2-(2,2-dimethylpropanamido)phenyl]boronic acid
Synonyms: 2-(2,2-dimethylpropanamido)phenylboronic acid
Molecular Weight (g/mol): 221.06
Molecular Formula: C11H16BNO3
InChi Key: MXRAJVMTCAUABO-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)NC1=CC=CC=C1B(O)O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results