accessibility menu, dialog, popup

UserName

CAS RN 146137-80-6

CH3OF

CAS RN 146137-80-6

IUPAC Name: 2-fluoro-4-methylbenzaldehyde
Synonyms: 2-fluoro-4-methylbenzaldehyde
Molecular Weight (g/mol): 138.14
Molecular Formula: C8H7FO
InChi Key: MVDRIMBGRZBWPE-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=O)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results