accessibility menu, dialog, popup

UserName

CAS RN 14589-38-9

OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHNd

CAS RN 14589-38-9

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 450.64
Molecular Formula: C15H30NdO6
InChi Key: BQBDJJRPGVHTDT-UHFFFAOYSA-T
SMILES: [Nd].CC(=[OH+])CC(C)=[OH+].CC(=[OH+])CC(C)=[OH+].CC(=[OH+])CC(C)=[OH+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results