accessibility menu, dialog, popup

UserName

CAS RN 145858-50-0

HClClNNNHN

CAS RN 145858-50-0

IUPAC Name: hydrogen 6-[(3-chlorophenyl)(1H-imidazol-1-yl)methyl]-1H-1,3-benzodiazole chloride
Synonyms: liarozole
Molecular Weight (g/mol): 345.23
Molecular Formula: C17H14Cl2N4
InChi Key: OTSZCHORPMQCBZ-UHFFFAOYNA-N
SMILES: [H+].[Cl-].ClC1=CC=CC(=C1)C(N1C=CN=C1)C1=CC=C2N=CNC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results