accessibility menu, dialog, popup

UserName

H3COOFFFO

CAS RN 145742-65-0

IUPAC Name: 2-methoxy-5-(trifluoromethoxy)benzaldehyde
Synonyms: 2-methoxy-5-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 220.15
Molecular Formula: C9H7F3O3
InChi Key: ATRDCTRZAJKDPL-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC(F)(F)F)C=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results