accessibility menu, dialog, popup

UserName

CAS RN 1450-93-7

NH2NHNOHSOHOONH2NHN

CAS RN 1450-93-7

IUPAC Name: 1H-imidazol-2-amine; sulfuric acid
Synonyms: 1H-imidazol-2-amine; sulfuric acid
Molecular Weight (g/mol): 181.17
Molecular Formula: C3H7N3O4S
InChi Key: LEUJVEZIEALICS-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=NC=CN1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)