CAS RN 1449248-72-9
CAS RN 1449248-72-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2’’-Desethyl-2’’-vinyl Simvastatin, TRC
CAS: 1449248-72-9 Molecular Formula: C25 H36 O5 Molecular Weight (g/mol): 416.55 Synonym: Methylene Simvastatin,(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2,2-Dimethylbut-3-enoate IUPAC Name: [(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] 2,2-dimethylbut-3-enoate SMILES: C[C@@H]1C[C@H](OC(=O)C(C)(C)C=C)[C@@H]2[C@@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H](C)C=CC2=C1