accessibility menu, dialog, popup

UserName

BONH3CCH3ClHOOH

CAS RN 1449144-75-5

IUPAC Name: [2-chloro-5-(dimethylcarbamoyl)phenyl]boronic acid
Synonyms: 2-chloro-5-(dimethylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 227.45
Molecular Formula: C9H11BClNO3
InChi Key: LVOHUMGEBQAIBQ-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C1=CC(B(O)O)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results