CAS RN 14400-67-0
IUPAC Name:
2,5-dimethyl-2,3-dihydrofuran-3-one
Synonyms:
2,5-dimethyl-2H-furan-3-one
Molecular Weight (g/mol):
112.13
Molecular Formula:
C6H8O2
InChi Key:
ASOSVCXGWPDUGN-UHFFFAOYNA-N
SMILES:
CC1OC(C)=CC1=O
1
–
1
of
1
results
1
2,5-Dimethyl-3(2H)-furanone, 94%
CAS: 14400-67-0 Molecular Formula: C6H8O2 Molecular Weight (g/mol): 112.128 MDL Number: MFCD00052571 InChI Key: ASOSVCXGWPDUGN-UHFFFAOYSA-N Synonym: 2,5-dimethyl-3 2h-furanone,2,5-dimethylfuran-3 2h-one,2,5-dimethyl-2,3-dihydrofuran-3-one,3 2h-furanone, 2,5-dimethyl,2,5-dimethyl-3 2h furanone,2,5-dimethyl-2h-furan-3-one,2,5-dimethyl-3-2h-furanone,2h-furan-3-one, 2,5-dimethyl,mango furanone PubChem CID: 85730 IUPAC Name: 2,5-dimethylfuran-3-one SMILES: CC1C(=O)C=C(O1)C