CAS RN 143322-55-8
CAS RN 143322-55-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)- 3-[(1-Methyl-2-pyrrolidinyl)methyl]-1H-Indole, TRC
CAS: 143322-55-8 Molecular Formula: C14H18N2 Molecular Weight (g/mol): 214.31 Synonym: 3-[[(2R)-1-Methyl-2-pyrrolidinyl]methyl]-1H-Indole IUPAC Name: 3-[[(2R)-1-methylpyrrolidin-2-yl]methyl]-1H-indole SMILES: CN1CCC[C@@H]1Cc2c[nH]c3ccccc23