accessibility menu, dialog, popup

UserName

CAS RN 143314-14-1

CH3NNH3CNOOO

CAS RN 143314-14-1

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium nitrate
Synonyms: 1-ethyl-3-methylimidazolium nitrate
Molecular Weight (g/mol): 173.17
Molecular Formula: C6H11N3O3
InChi Key: JDOJFSVGXRJFLL-UHFFFAOYSA-N
SMILES: [O-][N+]([O-])=O.CCN1C=C[N+](C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results