CAS RN 14320-38-8
CAS RN 14320-38-8
IUPAC Name:
cyclopent-3-en-1-ol
Synonyms:
cyclopent-3-en-1-ol
Molecular Weight (g/mol):
84.12
Molecular Formula:
C5H8O
InChi Key:
WEIMJSIRDZDHAH-UHFFFAOYSA-N
SMILES:
OC1CC=CC1
1
–
1
of
1
results
1
3-Cyclopenten-1-ol
CAS: 14320-38-8 Molecular Formula: C5H8O Molecular Weight (g/mol): 84.12 MDL Number: MFCD00798186 InChI Key: WEIMJSIRDZDHAH-UHFFFAOYSA-N IUPAC Name: cyclopent-3-en-1-ol SMILES: OC1CC=CC1