accessibility menu, dialog, popup

UserName

CH3OOHFFFOCH3

CAS RN 1431329-61-1

IUPAC Name: 4-methoxy-3-methyl-5-(trifluoromethyl)benzoic acid
Synonyms: 4-methoxy-3-methyl-5-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 234.17
Molecular Formula: C10H9F3O3
InChi Key: QUPPMFMWSDFHRQ-UHFFFAOYSA-N
SMILES: COC1=C(C)C=C(C=C1C(F)(F)F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results