accessibility menu, dialog, popup

UserName

CH3FOOHCl

CAS RN 1427416-28-1

IUPAC Name: 2-chloro-5-fluoro-3-methylbenzoic acid
Synonyms: 2-chloro-5-fluoro-3-methylbenzoic acid
Molecular Weight (g/mol): 188.58
Molecular Formula: C8H6ClFO2
InChi Key: AILHHXAQZADVIH-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C(=CC(F)=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results