accessibility menu, dialog, popup

UserName

CAS RN 141776-91-2

FFBr

CAS RN 141776-91-2

IUPAC Name: 1-(bromomethyl)-3,5-difluorobenzene
Synonyms: 1-(bromomethyl)-3,5-difluorobenzene
Molecular Weight (g/mol): 207.02
Molecular Formula: C7H5BrF2
InChi Key: KVSVNRFSKRFPIL-UHFFFAOYSA-N
SMILES: FC1=CC(CBr)=CC(F)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results