accessibility menu, dialog, popup

UserName

HONHONOHOH

CAS RN 140-07-8

IUPAC Name: 2-({2-[bis(2-hydroxyethyl)amino]ethyl}(2-hydroxyethyl)amino)ethan-1-ol
Synonyms: tetrahydroxyethyl ethylenediamine
Molecular Weight (g/mol): 236.31
Molecular Formula: C10H24N2O4
InChi Key: BYACHAOCSIPLCM-UHFFFAOYSA-N
SMILES: OCCN(CCO)CCN(CCO)CCO
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results