accessibility menu, dialog, popup

UserName

CAS RN 139705-74-1

H2NNN

CAS RN 139705-74-1

IUPAC Name: 4-{imidazo[1,2-a]pyridin-2-yl}aniline
Synonyms: 4-{imidazo[1,2-a]pyridin-2-yl}aniline
Molecular Weight (g/mol): 209.25
Molecular Formula: C13H11N3
InChi Key: FPCHINCJTRAFAK-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=CN2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results