accessibility menu, dialog, popup

UserName

OONH(S)H2NOOHHCl

CAS RN 139262-23-0

IUPAC Name: 6-amino-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid hydrochloride
Synonyms: 6-amino-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid hydrochloride
Molecular Weight (g/mol): 404.89
Molecular Formula: C21H25ClN2O4
InChi Key: MVMZFAIUUXYFGY-UHFFFAOYNA-N
SMILES: Cl.NCCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results