CAS RN 1391053-78-3
CAS RN 1391053-78-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Desfluoro Fluvoxketone, TRC
CAS: 1391053-78-3 Molecular Formula: C13H16F2O2 Molecular Weight (g/mol): 242.26 Synonym: (4-Methoxybutyl)(4-difluoromethylphenyl)methanone,Desfluoro EP Impurity D 5-Methoxy-1-[4-(difluoromethyl)phenyl]-1-pentanone SMILES: COCCCCC(=O)C1=CC=C(C=C1)C(F)F