CAS RN 138752-26-8
CAS RN 138752-26-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fellutamide B (Mixture of Diastereomers, >80%), TRC
CAS: 138752-26-8 Molecular Formula: C27H49N5O7 Molecular Weight (g/mol): 555.71 Synonym: N2-[(3R)-3-Hydroxy-1-oxododecyl]-L-asparaginyl-N1-(1-formyl-3-methylbutyl)-L-glutamamide,(2S)-2-((S)-4-Amino-2-((R)-3-hydroxydodecanamido)-4-oxobutanamido)-N1-(4-methyl-1-oxopentan-2-yl)pentanediamide IUPAC Name: (2S)-2-[[(2S)-4-amino-2-[[(3R)-3-hydroxydodecanoyl]amino]-4-oxobutanoyl]amino]-N-(4-methyl-1-oxopentan-2-yl)pentanediamide SMILES: CCCCCCCCC[C@@H](O)CC(=O)N[C@@H](CC(=O)N)C(=O)N[C@@H](CCC(=O)N)C(=O)NC(CC(C)C)C=O