accessibility menu, dialog, popup

UserName

HO(S)(R)OH(S)O(S)(S)HOOOOHNCl

CAS RN 138182-20-4

IUPAC Name: (2S,3S,4S,5R,6S)-6-[(6-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
Synonyms: (2S,3S,4S,5R,6S)-6-[(6-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
Molecular Weight (g/mol): 342.71
Molecular Formula: C14H13ClNO7
InChi Key: UFBPRKNLSYGHJJ-BYNIDDHOSA-M
SMILES: O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC(Cl)=CC=C23)O[C@@H]([C@H]1O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results