accessibility menu, dialog, popup

UserName

CAS RN 13814-97-6

SnFBFFFFBFFF

CAS RN 13814-97-6

IUPAC Name: λ²-tin(2+) bis(tetrafluoroboranuide)
Synonyms: λ²-tin(2+) ditetrafluoroborate
Molecular Weight (g/mol): 292.32
Molecular Formula: B2F8Sn
InChi Key: JPZSNSDMGTYJIW-UHFFFAOYSA-N
SMILES: [Sn++].F[B-](F)(F)F.F[B-](F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results