CAS RN 137975-06-5
CAS RN 137975-06-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Mozavaptan, TRC
CAS: 137975-06-5 Molecular Formula: C27H29N3O2 Molecular Weight (g/mol): 427.54 Synonym: N-[4-[[5-(Dimethylamino)-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl]carbonyl]phenyl]-2-methylbenzamide,OPC 31260; IUPAC Name: N-[4-[5-(dimethylamino)-2,3,4,5-tetrahydro-1-benzazepine-1-carbonyl]phenyl]-2-methyl-benzamide SMILES: CN(C)C1CCCN(C(=O)c2ccc(NC(=O)c3ccccc3C)cc2)c4ccccc14