accessibility menu, dialog, popup

UserName

OHCl

CAS RN 137736-28-8

IUPAC Name: [4-(4-chlorophenyl)cyclohexyl]methanol
Synonyms: [4-(4-chlorophenyl)cyclohexyl]methanol
Molecular Weight (g/mol): 224.73
Molecular Formula: C13H17ClO
InChi Key: INSWJXJRICZMOG-UHFFFAOYSA-N
SMILES: OCC1CCC(CC1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results