accessibility menu, dialog, popup

UserName

CAS RN 13765-25-8

IUPAC Name: europium(3+) trifluoride
Synonyms: europium(3+) trifluoride
Molecular Weight (g/mol): 208.96
Molecular Formula: EuF3
InChi Key: HPNURIVGONRLQI-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Eu+3]
Percent Purity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (3)
  • (2)
  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (5)