accessibility menu, dialog, popup

UserName

CAS RN 13737-36-5

OOHBr

CAS RN 13737-36-5

IUPAC Name: 2-[4-(bromomethyl)phenyl]acetic acid
Synonyms: [4-(bromomethyl)phenyl]acetic acid
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: WCOCCXZFEJGHTC-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(CBr)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)